Anti-Dopamine D₅ Receptor Rabbit Polyclonal Antibody

Supplier: MilliporeSigma
324408-100UL
EM324408-100ULEA 538.61 USD
EM324408-100UL
Anti-Dopamine D₅ Receptor Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Dopamine D₅ Receptor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR