Anti-Collagen Type X Rabbit Polyclonal Antibody

Supplier: MilliporeSigma
234196-500UL
EM234196-500ULEA 839.09 USD
EM234196-500UL
Anti-Collagen Type X Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Collagen Type X
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR