Anti-Cholera Toxin Goat Polyclonal Antibody

Supplier: MilliporeSigma
227040-100UL
EM227040-100ULEA 244.51 USD
EM227040-100UL
Anti-Cholera Toxin Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Cholera Toxin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR