Trichoderma viride Cellulase
Supplier: MilliporeSigma
219466-100KU 219466-200KU 219466-50KU
EM219466-100KUEA 311.54 USD
EM219466-100KU EM219466-200KU EM219466-50KU
Trichoderma viride Cellulase
Enzymes
From Trichoderma viride This enzyme shows a very high filter paper decomposing activity, as well as, appreciable xylanase and hemicellulase activity.
It is an effective enzyme to isolate intact cells and protoplasts from plant tissues.