5-Bromo-4-chloro-3-indolyl-α-D-glucopyranoside ≥97% (by HPLC), Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma

203781-25MG
EM203781-25MGEA 207.12 USD
EM203781-25MG
5-Bromo-4-chloro-3-indolyl-α-D-glucopyranoside ≥97% (by HPLC), Calbiochem®, Sigma-Aldrich®
5-Bromo-4-chloro-3-indolyl-α-D-glucopyranoside
Formula: C₁₄H₁₅BrClNO₆
MW: 408.63 g/mol
Storage Temperature: Freezer
CAS Number: 1174456-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR