D-Biotin-N-succinimidyl ester ≥90% (by NMR), Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Biotin-NHS, 2,5-Dioxopyrrolidin-1-yl 5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoate, (+)-Biotin N-hydroxysuccinimide ester, D-Biotinyl-OSu, NHS-Biotin, Biotin N-hydroxysuccinimide ester, N-(D-Biotinyloxy)succinimide

203112-250MG
EM203112-250MGEA 486.63 USD
EM203112-250MG
D-Biotin-N-succinimidyl ester ≥90% (by NMR), Calbiochem®, Sigma-Aldrich®
D-Biotin-N-succinimidyl ester
Formula: C₁₄H₁₉N₃O₅S
Storage Temperature: Refrigerator
MDL Number: MFCD00078531
CAS Number: 35013-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR