D-(+)-Biotin ≥97% (by assay), Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma

2031-1GM
EM2031-1GMEA 136.39 USD
EM2031-1GM
D-(+)-Biotin ≥97% (by assay), Calbiochem®, Sigma-Aldrich®
D(+)-Biotin
Formula: C₁₀H₁₆N₂O₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00005541
CAS Number: 58-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR