ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulfonate]), Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) diammonium salt, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulfonate), ABTS, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulphonate), 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) diammonium salt

194430-1GM
EM194430-1GMEA 247.66 USD
EM194430-1GM
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulfonate]), Sigma-Aldrich®
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])

ABTS™ Chromophore, Diammonium Salt


Useful as a substrate for ELISA; acts as a hydrogen donor in peroxidase reactions. Radical cation has absorption maximum at 414 nm.

Formula: C₁₈H₂₄N₆O₆S₄
MW: 548.68 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00010404
CAS Number: 30931-67-0
EINECS: 250-396-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR