Anti-APOE Goat Polyclonal Antibody

Supplier: MilliporeSigma
178479-500UL
EM178479-500ULEA 616.97 USD
EM178479-500UL
Anti-APOE Goat Polyclonal Antibody
Antibodies
Type:
Antigen: Apolipoprotein E
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR