Anti-Apolipoprotein B Goat Polyclonal Antibody

Supplier: MilliporeSigma
178467-1ML
EM178467-1MLEA 178.14 USD
EM178467-1ML
Anti-Apolipoprotein B Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Apolipoprotein B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR