Agar, Calbiochem®

Supplier: MilliporeSigma

Synonyms: Agar Agar

12177-1KG
EM12177-1KGEA 354.58 USD
EM12177-1KG
Agar, Calbiochem®
Agar

Suitable for inclusion in culture media.


Storage and Shipping Instructions: Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 1 month at 4°C.

Melting Pt: 90
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
Merck Index: 14, 184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form Beige solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR