Water ≥99.9% tested for EPA533 and EPA537.1 PFAS Methods, LiChrosolv®, Supelco®

Supplier: MilliporeSigma

1047351000
EM1047351000EA 91.1 USD
EM1047351000
Water ≥99.9% tested for EPA533 and EPA537.1 PFAS Methods, LiChrosolv®, Supelco®
Water
Formula: H₂O
Storage Temperature: Room Temperature
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Specification Test Results

EPA 533/537.1 Tested Conforms
UHPLC Gradient (at 210 nm) ≤ 2 mAU
Evaporation Residue ≤ 0.7 mg/l
UHPLC Gradient (at 254 nm) ≤ 0.5 mAU
(PEG) Signal-to-Noise Ratio ≤50


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR