Acetonitrile ≥99.9% tested for EPA533 and EPA537.1 PFAS Methods, LiChrosolv®, Supelco®

Supplier: MilliporeSigma
Danger

1047261000
EM1047261000EA 352.9 USD
EM1047261000
Acetonitrile ≥99.9% tested for EPA533 and EPA537.1 PFAS Methods, LiChrosolv®, Supelco®
Acetonitrile
Formula: C₂H₃N
Storage Temperature: Room Temperature
MDL Number: MFCD00001878
CAS Number: 75-05-8
UN: 1648
REACH: 01-2119471307-38-XXXX

Order Now

Specification Test Results

EPA 533/537.1 Tested Conforms
UHPLC Gradient (at 210 nm) ≤1.0 mAU
Evaporation Residue ≤ 0.7 mg/l
Titratable Acid (meq/g) ≤ 0.001
Titratable Base (meq/g) ≤ 0.0001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR