Buffer Solution, Blue, Traceable to NIST and PTB pH 10.00 (25 °C), Certipur®, MilliporeSigma
Supplier: MilliporeSigma
1.99050.4000
EM1.99050.4000EA
245
USD
EM1.99050.4000
Buffer Solution, Blue, Traceable to NIST and PTB pH 10.00 (25 °C), Certipur®, MilliporeSigma
Buffers
pH Calibration Buffers
Buffer solution (boric acid/potassium chloride/sodium hydroxide) colour coded: blue, traceable to NIST and PTB pH 10.00 (25°C) Certipur®.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...