Supelco Spectroquant® Move DC Colorimeter, Merck
Supplier: MilliporeSigma
1.73635.0001
EM1.73635.0001EA 894.15 USD
EM1.73635.0001
Supelco Spectroquant® Move DC Colorimeter, Merck
Colorimeters Multiparameter Colorimeters
The Spectroquant® Move Cl2/O3/ClO2/CyA/pH mobile colorimeter is suitable for the detection of five essential parameters of water analysis: chlorine, ozone, chlorine dioxide, cyanuric acid and pH
This handy, battery-operated instrument comes with a carrying case and is designed for in-process and field work.
Ordering information: Standard contents of the Spectroquant Colorimeter Move 100 include the colorimeter in a plastic carrying case, 4 batteries (type AA/LR6), adapter for 16mm dia. round cells, 3 round cells with 16mm dia. cap, 3 round cells with 24mm dia. cap, screwdriver, operating instructions manual, and certificate of compliance.