Titanium Standard for ICP, Certipur®, MilliporeSigma
Supplier: MilliporeSigma
1.70363.0100
EM1.70363.0100CS 5005 USD
EM1.70363.0100
Titanium Standard for ICP, Certipur®, MilliporeSigma
Standards Cationic Standards Titanium Standards
Traceable to SRM from NIST.
Certifications: The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
Ordering information: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.