Scandium Standard for ICP, Certipur®, MilliporeSigma
Supplier: MilliporeSigma
1.70349.0100
EM1.70349.0100EA
211
USD
EM1.70349.0100
Scandium Standard for ICP, Certipur®, MilliporeSigma
Standards
Cationic Standards
Scandium Standards
Traceable to SRM from NIST.
Certifications: The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
Ordering information: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)