Platinum Standard for AAS, Certipur®, MilliporeSigma
Supplier: MilliporeSigma
1.70219.0500
EM1.70219.0500CS 5197 USD
EM1.70219.0500
Platinum Standard for AAS, Certipur®, MilliporeSigma
Standards Cationic Standards Platinum Standards
H₂PtCl₆ in HCl
Traceable to SRM from NIST.