Cesium Standard for AAS, Certipur®, MilliporeSigma
Supplier: MilliporeSigma
1.70212.0100
EM1.70212.0100CS
4042
USD
EM1.70212.0100
Cesium Standard for AAS, Certipur®, MilliporeSigma
Standards
Cationic Standards
Cesium Standards
Traceable to SRM from NIST.