Oxalic acid c(CsH2O4) = 0.005 mol; standardised against Potassium permangante solution, Titripur®, Supelco®

Supplier: MilliporeSigma

Synonyms: Ethanedioic acid

1.60329.1000
EM1.60329.1000EA 77 USD
EM1.60329.1000
Oxalic acid c(CsH2O4) = 0.005 mol; standardised against Potassium permangante solution, Titripur®, Supelco®
Oxalic acid
Formula: C₂H₂O₄
MW: 90.04 g/mol
Boiling Pt: 365
MDL Number: MFCD00002573
CAS Number: 144-62-7
Merck Index: 13,06980

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR