(±)-α-Methoxyphenylacetic acid, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

1.59712.0010
EM1.59712.0010EA 486.03 USD
EM1.59712.0010
(±)-α-Methoxyphenylacetic acid, Supelco®
(±)-α-Methoxyphenylacetic acid
Formula: C₉H₁₀O₃
Boiling Pt: 165 °C (24 hPa)
Melting Pt: 69…71 °C
MDL Number: MFCD00064217
CAS Number: 7021-09-2

Order Now

Specification Test Results

Appearance White to almost white crystals or crystalline powder.
Identity (IR-spectrum) conforms
Melting point 68 - 72 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR