Ammonium carbonate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
1.59504.0250
1.59504.1000
EM1.59504.0250CS
375
USD
EM1.59504.0250
EM1.59504.1000
Ammonium carbonate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Specification Test Results
Assay (acidimetric, NH₃) | ≥ 30.0 % |
Identity | passes test |
Description | white crystals |
Insoluble matter | ≤ 0.005 % |
Chloride (Cl) | ≤ 0.0005 % |
Sulfur compounds (as SO₄) | ≤ 0.002 % |
Heavy metals (as Pb) | ≤ 0.0005 % |
Fe (Iron) | ≤ 0.0005 % |
Non-volatile matter | ≤ 0.01 % |
Conforms ACS, Reag. Ph. Eur. | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...