Metanil yellow, Millipore®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

1.59267.0010
EM1.59267.0010CS 4925 USD
EM1.59267.0010
Metanil yellow, Millipore®
Metanil Yellow
Formula: C₁₈H₁₄N₃NaO₃S
MDL Number: MFCD00007486
CAS Number: 587-98-4
EINECS: 209-608-2

Order Now

Specification Test Results

Solubility 25 g/l (20 °C)
Molar Mass 375.38 g/mol
Bulk Density 330 kg/m3
pH 10 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR