Acetonitrile with 0.1% (v/v) trifluoroacetic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ100

1.59014.2500
EM1.59014.2500EA 287 USD
EM1.59014.2500
Acetonitrile with 0.1% (v/v) trifluoroacetic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®
Acetonitrile with 0.1% (v/v) Trifluoroacetic acid
Density: 0.786 g/cm3 (20 °C)
Flash Pt: 2 °C
Storage Temperature: Ambient
UN: 1648
ADR: 3,

Order Now

Specification Test Results

Acid content 0.095 - 0.105 % (v/v)
Identity passes test
Color ≤ 10 Hazen
Al (Aluminium) ≤ 0.05 ppm
Ca (Calcium) ≤ 0.05 ppm
Fe (Iron) ≤ 0.05 ppm
K (Potassium) ≤ 0.025 ppm
Mg (Magnesium) ≤ 0.05 ppm
Na (Sodium) ≤ 0.25 ppm
Gradient grade (at 210 nm) ≤ 1.0 mAU
Gradient grade (at 254 nm) ≤ 0.5 mAU
Suitable for LC-MS; Intensity of background mass peak based on reserpine (APCI/ESI positive) ≤ 2 ppb
Filtered by 0.2 µm filter.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR