Water with 0,1% (v/v) formic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ100
1.59013.2500
EM1.59013.2500EA 106 USD
EM1.59013.2500
Water with 0,1% (v/v) formic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®
Water with 0,1% (v/v) formic acid
LiChrosolv® hypergrade is accurately tested for LC-MS suitability, and meet all the requirements of modern LC-MS ionization methods (ESI/APCI - positive and negative mode).
Thanks to their low level of ionic background and low ion suppression, LiChrosolv® hypergrade solvents provide high reproducibility and high ionization efficiency. The packaging material has been improved to meet LC-MS quality requirements perfectly.