Acetonitrile with 0.1% (v/v) Formic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ100

1.59002.1000 1.59002.2500
EM1.59002.1000EA 191 USD
EM1.59002.1000 EM1.59002.2500
Acetonitrile with 0.1% (v/v) Formic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®
Acetonitrile with 0,1% (v/v) Formic acid

LiChrosolv® hypergrade is accurately tested for LC-MS suitability, and meet all the requirements of modern LC-MS ionization methods (ESI/APCI - positive and negative mode).


Thanks to their low level of ionic background and low ion suppression, LiChrosolv® hypergrade solvents provide high reproducibility and high ionization efficiency. The packaging material has been improved to meet LC-MS quality requirements perfectly.

Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: −45,7 °C
Flash Pt: 2 °C
Storage Temperature: Ambient
UN: 1648
ADR: 3,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR