Calcium chloride dihydrate, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, FCC, E509, SAFC®

Supplier: MilliporeSigma
Warning

1.42002.9029 1.42002.1000 1.42002.5000 1.42002.9525
EM1.42002.9029EA 785 USD
EM1.42002.9029 EM1.42002.1000 EM1.42002.5000 EM1.42002.9525
Calcium chloride dihydrate, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, FCC, E509, SAFC®
Calcium chloride dihydrate

pH value: 4.5 to 8.5 (50 g/L, H₂O, 20 °C)
Solubility: 1280 g/L

Formula: CaCl₂·2H₂O
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 176 °C
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR