Sodium hydroxide 0.1 mol/L in aqueous solution, EMPROVE® EXPERT for biopharmaceutical production, SAFC®

Supplier: MilliporeSigma

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

Filtered through a 0.2 µm filter.

1.37058.9025 1.37058.9950
EM1.37058.9025EA 432.83 USD
EM1.37058.9025 EM1.37058.9950
Sodium hydroxide 0.1 mol/L in aqueous solution, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Sodium hydroxide

Suitable for biopharmaceutical production.


This product has been manufactured from Sodium hydroxide pellets Ph. Eur., BP, JP, NF and waterfor injection (wfi) ace. USP which conforms with highly purified water (hpw) ace. Ph Eur.


Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elementallmpurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Formula: NaOH
Density: 1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,

Order Now

Specification Test Results

Assay (acidimetric,NaOH) 0.05- 0.15 mol/l
Carbonate (as Na2CO3) ≤ 0.2%
Chloride (Cl) ≤ 5%
Sulfate (SO.) ≤ 30 ppm
Heavy metals (as Pb) ≤ 5.0 ppm
Al (Aluminium) ≤ 10 ppm
Fe (Iron) ≤ 3 ppm
Residual solvents (ICH Q3C) excluded by production process
Endotoxins ≤ 2.0 I.U./ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR