Sodium hydroxide (5 - 60%) in aqueous solution solution 32% (by acidimetry), EMPROVE® EXPERT for biopharmaceutical production, SAFC®

Supplier: MilliporeSigma
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

Filtered through a 0.2 µm filter.

1.37023.5000 1.37023.9025 1.37023.9200 1.37023.2500
EM1.37023.5000CS 1938.14 USD
EM1.37023.5000 EM1.37023.9025 EM1.37023.9200 EM1.37023.2500
Sodium hydroxide (5 - 60%) in aqueous solution solution 32% (by acidimetry), EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Sodium hydroxide

This product has been manufactured from Sodium hydroxide pellets Ph Eur, BP, JP, NF and water for injection (wfi) acc. USP which conforms with highly purified water (hpw) acc. Ph Eur.


Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Formula: NaOH
Melting Pt: 9 °C
Density: 1.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,

Order Now

Specification Test Results

Assay (acidimetric,NaOH) 32.0 - 33.0 %
Carbonate (as Na₂CO₃) ≤ 1.0 %
Chloride (Cl) ≤ 5 ppm
Sulfate (SO₄) ≤ 30 ppm
Heavy metals (as Pb) ≤ 5.0 ppm
Al (Aluminium) ≤ 10 ppm
Fe (Iron) ≤ 3 ppm
Ni (Nickel) * ≤ 2.00 ppm
Residual solvents (ICH Q3C) ...excluded by production process
Endotoxins ≤ 2.0 I.U./ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR