Phosphorus (total) Standard Solution, CRM traceable to SRM from NIST 0.400 mg/L PO4-P (total) in H₂O
Supplier: MilliporeSigma
1.25046.0100
EM1.25046.0100CS 2851 USD
EM1.25046.0100
Phosphorus (total) Standard Solution, CRM traceable to SRM from NIST 0.400 mg/L PO4-P (total) in H₂O
Standards Cationic Standards Phosphorus Standards