Silver Standard for AAS, Certipur®, MilliporeSigma
Supplier: MilliporeSigma
1.19797.0100
EM1.19797.0100EA 79 USD
EM1.19797.0100
Silver Standard for AAS, Certipur®, MilliporeSigma
Standards Cationic Standards Silver Standards
Traceable to SRM from NIST.