Dipotassium hydrogen orthophosphate trihydrate, LiChropur® buffer substance, for chromatography, Supelco®

Supplier: MilliporeSigma

Synonyms: Dipotassium hydrogen phosphate trihydrate, Potassium hydrogen phosphate trihydrate, di-Potassium hydrogen phosphate trihydrate, Potassium phosphate dibasic trihydrate, di-Potassium hydrogen orthophosphate trihydrate

M-Clarity™ quality level = MQ100

1.19754.0250
EM1.19754.0250CS 2599 USD
EM1.19754.0250
Dipotassium hydrogen orthophosphate trihydrate, LiChropur® buffer substance, for chromatography, Supelco®
di-Potassium hydrogen orthophosphate trihydrate
Formula: K₂HPO₄·3H₂O
MW: 228.22 g/mol
Boiling Pt: ∼340 °C (1013 hPa)
Density: 2.504 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149926
CAS Number: 16788-57-1
EINECS: 231-834-5

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 340 °C (anhydrous substance),decomposes
Molar Mass 228.23 g/mol
Bulk Density 800 kg/m3
pH 9.2 - 9.4 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR