Disodium hydrogen phosphate dihydrate ≥99.0% (by acidimetry), LiChropur™ buffer substance, for chromatography, Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ100

1.19753.0250
EM1.19753.0250CS 1910.1 USD
EM1.19753.0250
Disodium hydrogen phosphate dihydrate ≥99.0% (by acidimetry), LiChropur™ buffer substance, for chromatography, Supelco®
di-Sodium hydrogen phosphate dihydrate
Formula: Na₂HPO₄·2H₂O
Melting Pt: 92.5 °C
Density: 2.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149182
CAS Number: 10028-24-7
EINECS: 231-448-7

Order Now

Specification Test Results

Identity passes test
pH-value (5 %; water) 9.0 - 9.2
Assay (acidimetric) ≥ 99.0 %
Self-absorption (215 nm; 0.2 mol/l; 1 cm; water) ≤ 0.15
Self-absorption (220 nm; 0.2 mol/l; 1 cm; water) ≤ 0.05
Self-absorption (254 nm; 0.2 mol/l; 1 cm; water) ≤ 0.02


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR