Oxygen Scavengers Test, Spectroquant®, MilliporeSigma
Supplier: MilliporeSigma
1.19251.0001
EM1.19251.0001EA
297
USD
EM1.19251.0001
Oxygen Scavengers Test, Spectroquant®, MilliporeSigma
Environmental Test Kits
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)