Platinum, black ≥98%, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Platinium

M-Clarity™ quality level = MQ300

1.19233.0005
EM1.19233.0005CS 98543 USD
EM1.19233.0005
Platinum, black ≥98%, Supelco®
Platinum
Formula: Pt
MW: 195.08 g/mol
MDL Number: MFCD00011179
CAS Number: 7440-06-4
UN: 1479
ADR: 5.1,

Order Now

Specification Test Results

Melting Point 1770 °C
Molar Mass 195.08 g/mol
Density 21.45 g/cm3
pH (H2O) not applicable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR