MColortest™ Phosphate Test, MilliporeSigma
Supplier: MilliporeSigma
1.18388.0001 1.18466.0002
EM1.18388.0001EA 416 USD
EM1.18388.0001 EM1.18466.0002
MColortest™ Phosphate Test, MilliporeSigma
Environmental Test Kits
The test can be used for the determination of orthophosphate in groundwater and surface water, drinking water, wastewater
When the reagents of the MColortestTM test are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of MColortestTM test kits, making it a particularly economical system.
The concentration can be measured semiquantitatively by using the colour comparator.