1-Dodecansulfonic acid sodium salt, Supelco®

Supplier: MilliporeSigma

Synonyms: Dodecane-1-sulfonic acid sodium salt, Sodium 1-dodecanesulphonate, Dodecane-1-sulphonic acid sodium salt, Sodium 1-dodecansulphonate, 1-Dodecanesulphonic acid, sodium salt, 1-Dodecanesulfonic acid, sodium salt, Sodium dodecane-1-sulphonate, Sodium dodecane-1-sulfonate, Sodium 1-dodecansulfonate, Sodium 1-dodecanesulfonate

M-Clarity™ quality level = MQ100

1.18308.0025
EM1.18308.0025CS 13021.71 USD
EM1.18308.0025
1-Dodecansulfonic acid sodium salt, Supelco®
1-Dodecansulphonic acid sodium salt

Dodecane-1-sulfonic acid sodium salt for ion pair chromatography LiChropur®

Formula: C₁₂H₂₅NaO₃S
MW: 272.38 g/mol
MDL Number: MFCD00007527
CAS Number: 2386-53-0

Order Now

Specification Test Results

Hill Formula C₁₂H₂₅NaO₃S
Molar Mass 272.37 g/mol
pH value 5.5 - 7.5 (100 g/l, H₂O, 20 °C)
Bulk density 330 kg/m³
Solubility (20 °C) soluble
Storage class 10 - 13 Other liquids and solids
WGK WGK 2 water endangering

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR