1-Octanesulfonic acid sodium salt, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Sodium 1-octansulfonate, Sodium octane-1-sulphonate

M-Clarity™ quality level = MQ100

1.18307.0025
EM1.18307.0025CS 13021.71 USD
EM1.18307.0025
1-Octanesulfonic acid sodium salt, Supelco®
1-Octanesulphonic acid sodium salt

Octane-1-sulfonic acid sodium salt for ion pair chromatography LiChropur®.

Formula: C₈H₁₇NaO₃S
MW: 216.28 g/mol
Melting Pt: 300 °C
MDL Number: MFCD00007544
CAS Number: 5324-84-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1244cb11db07775dec2e193823e40080024b7036&vid=32d95cd9e321818e2f94c8261c4b4e6180ee0fc7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (alkalimetric, calculated on dried substance) ≥ 99 %
Identity (IR-spectrum) conforms
pH (100 g/l, water) 5.5 - 7.5
UV-transmission (0.005 mol/l; 1 cm; Water) (at 200 nm) ≥ 70 %
UV-transmission (0.005 mol/l; 1 cm; Water) (at 220 nm) ≥ 90 %
UV-transmission (0.005 mol/l; 1 cm; Water) (at 250 nm) ≥ 98 %
Loss on drying (120 °C; 4h; Vacuum) ≤ 2.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR