1-Heptanesulfonic acid sodium salt, LiChropur™ for ion pair chromatography, Supelco®

Supplier: MilliporeSigma

Synonyms: Sodium heptane-1-sulfonate, Sodium 1-heptanesulfonate, Sodium heptane-1-sulphonate, Sodium 1-heptanesulphonate

M-Clarity™ quality level = MQ100

1.18306.0025
EM1.18306.0025CS 13021.71 USD
EM1.18306.0025
1-Heptanesulfonic acid sodium salt, LiChropur™ for ion pair chromatography, Supelco®
1-Heptanesulphonic acid sodium salt

Heptane-1-sulfonic acid sodium salt for ion pair chromatography LiChropur®.

Formula: H₃C(CH₂)₅CH₂SO₃Na
MW: 202.25 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00007543
CAS Number: 22767-50-6
EINECS: 245-210-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95c09a5691140bab7446d88919f01593e8f0b9b2&vid=7c237cde5db01d0567ff19964ca3287434747480&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Hill Formula C₇H₁₅NaO₃S
Molar Mass 202.24 g/mol
Melting Point 300 °C
pH value 5.5 - 7.5 (100 g/l, H₂O, 20 °C)
Bulk density 391 kg/m³
Solubility (20 °C) soluble
Storage class 10 - 13 Other liquids and solids
WGK WGK 1 slightly water endangering

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR