1-Pentanesulfonic acid sodium salt, Supelco®

Supplier: MilliporeSigma

Synonyms: Sodium 1-pentanesulphonate, Sodium pentane-1-sulphonate, Pentane-1-sulphonic acid sodium salt, Sodium pentanesulphonate, 98%, Sodium pentanesulphonate, Pentane-1-sulfonic acid sodium salt, Sodium pentanesulfonate, Sodium 1-pentanesulfonate, Sodium pentane-1-sulfonate, Sodium pentanesulfonate, 98%

M-Clarity™ quality level = MQ100

1.18304.0025
EM1.18304.0025CS 13030.81 USD
EM1.18304.0025
1-Pentanesulfonic acid sodium salt, Supelco®
1-Pentanesulphonic acid sodium salt

Pentane-1-sulfonic acid sodium salt for ion pair chromatography LiChropur®.

Formula: C₅H₁₁O₃SNa
MW: 174.2 g/mol
MDL Number: MFCD00007541
CAS Number: 22767-49-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddb1c23c8a6fe3c156a3aacb62721e1d1a34efb0&vid=1936dcdb47ba374ce21fd758660f7ca6ba95d254&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Hill Formula C₅H₁₁NaO₃S
Molar Mass 174.20 g/mol
pH value 5.5 - 7.5 (100 g/l, H₂O, 20 °C)
Bulk density 390 kg/m³
Solubility (20 °C) soluble
Storage class 10 - 13 Other liquids and solids
WGK WGK 3 highly water endangering

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR