1-Butanesulfonic acid sodium salt, Supelco®

Supplier: MilliporeSigma

Synonyms: Sodium 1-butanesulphonate, Sodium butane-1-sulphonate, Sodium 1-butanesulfonate, Butane-1-sulfonic acid sodium salt, Sodium 1-butansulphonate, Sodium 1-butansulfonate, Butane-1-sulphonic acid sodium salt, Sodium butane-1-sulfonate

M-Clarity™ quality level = MQ100

1.18303.0025
EM1.18303.0025CS 9317.05 USD
EM1.18303.0025
1-Butanesulfonic acid sodium salt, Supelco®
1-Butansulphonic acid sodium salt

Butane-1-sulfonic acid sodium salt for ion pair chromatography LiChropur®.

Formula: C₄H₉NaO₃S
MW: 160.17 g/mol
MDL Number: MFCD00007540
CAS Number: 2386-54-1

Order Now

Specification Test Results

Hill Formula C₄H₉NaO₃S
Molar Mass 160.17 g/mol
Melting Point 310 °C
Storage class 10 - 13 Other liquids and solids
WGK WGK 3 highly water endangering

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR