Quaternary Ammonium Compounds Method: Colorimetric With Test Strips 10 - 25 - 50 - 100 - 250 - 500 mg/L MQuant™, MilliporeSigma
Supplier: MilliporeSigma
1.17920.0001
EM1.17920.0001CS 18426 USD
EM1.17920.0001
Quaternary Ammonium Compounds Method: Colorimetric With Test Strips 10 - 25 - 50 - 100 - 250 - 500 mg/L MQuant™, MilliporeSigma
Environmental Test Kits
The test strip can be used to check quaternary ammonium compounds, which have microbicidal properties and are mainly used in surface cleaners.