Arsenic Test Method: colorimetric with test strips and reagents 0.02 - 0.05 - 0.1 - 0.2 - 0.5 mg/L As 0.1 - 0.5 - 1.0 - 1.7 - 3.0 mg/L As MQuant™, MilliporeSigma
Supplier: MilliporeSigma
1.17917.0001
EM1.17917.0001CS 1877 USD
EM1.17917.0001
Arsenic Test Method: colorimetric with test strips and reagents 0.02 - 0.05 - 0.1 - 0.2 - 0.5 mg/L As 0.1 - 0.5 - 1.0 - 1.7 - 3.0 mg/L As MQuant™, MilliporeSigma
Environmental Test Kits