MQuant™ Free Fatty Acids Test, MilliporeSigma
Supplier: MilliporeSigma
1.17046.0001
EM1.17046.0001CS 18807 USD
EM1.17046.0001
MQuant™ Free Fatty Acids Test, MilliporeSigma
Food and Beverage Test Systems
MQuant™ free fatty acids colorimetric test, 0.5, 1, 2, 3 mg/g KOH.