Sulfite Test Method: Reflectometric With Test Strips 10 - 200 mg/L SO32- Reflectoquant®, MilliporeSigma
Supplier: MilliporeSigma
1.16987.0001
EM1.16987.0001CS 19269 USD
EM1.16987.0001
Sulfite Test Method: Reflectometric With Test Strips 10 - 200 mg/L SO32- Reflectoquant®, MilliporeSigma
Environmental Test Kits
Test strips for the determination of sulfite with reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Test kits with reagents for reliable documented quantitative results
  • Barcode calibration
  • Low analysis cost
  • Test strips with minute amount of chemicals for easy disposal
The test can be used for the determination of sulfite ions, which form a red compound in reaction with the reagents, in e.g. wastewater, boiler water and boiler feed water and food after appropriate sample pretreatment.