Ascorbic Acid Test, 25 - 450 mg/L, Reflectoquant®, MilliporeSigma
Supplier: MilliporeSigma
1.16981.0001
EM1.16981.0001CS 24261 USD
EM1.16981.0001
Ascorbic Acid Test, 25 - 450 mg/L, Reflectoquant®, MilliporeSigma
Environmental Test Kits
Ascorbic Acid (vitamin C) counts as an essential quality characteristic in many foods. Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
MilliporeSigma # 1.16981.0001 replaces and is identical to MilliporeSigma # 16981-1
The content of ascorbic acid is measured during the production and filling procedures in the fruit- and vegetable-processing sector as a measure to guarantee compliance of the minimum required values for preservation.