Nitrate Test Method: Reflectometric With Test Strips 5 - 225 mg/L NO₃⁻ Reflectoquant®, MilliporeSigma
Supplier: MilliporeSigma
1.16971.0001
EM1.16971.0001CS
24261
USD
EM1.16971.0001
Nitrate Test Method: Reflectometric With Test Strips 5 - 225 mg/L NO₃⁻ Reflectoquant®, MilliporeSigma
Environmental Test Kits
The rapid determination of the nitrate content is of particular interest with fruit and vegetables to help decide whether a given delivery batch can be released for use in a specific product (e.g baby foods).
MilliporeSigma # 1.16971.0001 replaces and is identical to MilliporeSigma # 16971-1
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.