Reflectoquant® pH Test in Cooling Lubricants, MilliporeSigma
Supplier: MilliporeSigma
1.16898.0001
EM1.16898.0001CS 11569 USD
EM1.16898.0001
Reflectoquant® pH Test in Cooling Lubricants, MilliporeSigma
Test Paper pH Indicator Papers and Test Strips
In water analysis the pH value plays a very important role in how other substances behave. The test can be used for monitoring the pH value in cooling lubricants.
  • Store at 15 to 25 °C
Reflectoquant® pH tests are suited for all media in environmental analysis and in industrial in-process controls. The reaction zones of the test strip change color depending on the pH. The responsible indicator is chemically bound to the zones and hence does not bleed. The resulting color is evaluated reflectometrically on the RQflex®.