Diethanolamine, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ300

1.16205.1000
EM1.16205.1000CS 680 USD
EM1.16205.1000
Diethanolamine, EMSURE® for analysis, Supelco®
Diethanolamine
Formula: C₄H₁₁NO₂
Boiling Pt: 268.8 °C (1013 hPa)
Melting Pt: 28 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 176 °C
Storage Temperature: Ambient
MDL Number: MFCD00002843
CAS Number: 111-42-2
EINECS: 203-868-0
REACH: 01-2119488930-28
Merck Index: 13,03134

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 28 °C
Molar Mass 105.14 g/mol
Boiling Point 269 - 270 °C (1013 hPa)
Vapor Pressure  - 0.01 hPa (25 °C)
Flash Point 176 °C
Refractive Index 1.4753 (30 °C, 589 nm)
Explosion Limit 2.1 - 10.6 %(V)
Density 1.09 g/cm3 (30 °C) liquid
pH 11 (53 g/l, H2O, 20 °C)
Ignition Point 365 - 370 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR