Acridine orange (hemizinc salt), Certistain® for microscopy (C.I. 46005), Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt, Euchrysine 3RX, Acridine orange hemi(Zinc chloride) salt, Acridine orange, Basic orange 14, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt), Euchrysin GNY, N,N,N′,N′-Tetramethylacridine-3,6-diamine monohydrochloride compd. with zinc dichloride

1.15931.0025
EM1.15931.0025CS 8418 USD
EM1.15931.0025
Acridine orange (hemizinc salt), Certistain® for microscopy (C.I. 46005), Sigma-Aldrich®
Acridine orange (hemizinc salt)
Formula: C₁₇H₂₀ClN₃·HCl·½ZnCl₂
MW: 369.96 g/mol
MDL Number: MFCD00081043
CAS Number: 10127-02-3

Order Now

Specification Test Results

Solubility 28 g/l (20 °C)
Melting Point 165 °C
Molar Mass 438.11 g/mol
Bulk Density 200 kg/m3
pH 3.9 (10 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR